Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015747.1 | NP_001015747.1 | 2 - 3 | 318 | 118 | |
XM_012966538.2 | XP_012821992.1 | 2 - 3 | 222 | 118 | ||
GenBank mRNA | BC089683.1 | - | 2 - 3 | 318 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |