Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015766.1 | NP_001015766.1 | 1 - 2 | 178 | 89 | |
GenBank mRNA | BC089705.1 | - | 1 - 2 | 178 | 89 | |
CU025071.1 | - | 1 - 2 | 142 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |