Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015766.1 | NP_001015766.1 | 2 - 3 | 361 | 82 | |
GenBank mRNA | BC089705.1 | - | 2 - 3 | 361 | 82 | |
CU025071.1 | - | 2 - 3 | 325 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |