Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfpi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015766.1 | NP_001015766.1 | 3 - 4 | 538 | 102 | |
GenBank mRNA | BC089705.1 | - | 3 - 4 | 538 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |