Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015779.1 | NP_001015779.1 | 3 - 4 | 419 | 79 | |
GenBank mRNA | BC089720.1 | - | 3 - 4 | 419 | 79 | |
CT030280.1 | - | 3 - 4 | 424 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |