Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp3a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015786.1 | NP_001015786.1 | 8 - 9 | 833 | 104 | |
GenBank mRNA | BC089731.1 | - | 8 - 9 | 833 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |