Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cd37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015801.2 | NP_001015801.2 | 6 - 7 | 540 | 94 | |
GenBank mRNA | BC089757.1 | - | 6 - 7 | 540 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |