Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015849.1 | NP_001015849.1 | 4 - 5 | 340 | 73 | |
XM_012961773.1 | XP_012817227.1 | 4 - 5 | 524 | 73 | ||
GenBank mRNA | BC090139.1 | - | 4 - 5 | 340 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |