Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015849.1 | NP_001015849.1 | 5 - 6 | 419 | 64 | |
XM_012961773.1 | XP_012817227.1 | 5 - 6 | 603 | 64 | ||
GenBank mRNA | BC090139.1 | - | 5 - 6 | 419 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |