Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ybx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015855.1 | NP_001015855.1 | 4 - 5 | 410 | 67 | |
XM_012966546.2 | XP_012822000.1 | 4 - 5 | 437 | 67 | ||
GenBank mRNA | BC090361.1 | - | 4 - 5 | 410 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |