Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015859.1 | NP_001015859.1 | 5 - 6 | 645 | 66 | |
XM_012966550.2 | XP_012822004.1 | 5 - 6 | 786 | 66 | ||
GenBank mRNA | BC090370.1 | - | 5 - 6 | 645 | 66 | |
CT027850.1 | - | 5 - 6 | 627 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |