Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hba3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015904.2 | NP_001015904.1 | 1 - 2 | 122 | 94 | |
GenBank mRNA | BC075607.1 | - | 1 - 2 | 120 | 94 | |
BC135231.1 | - | 1 - 2 | 136 | 94 | ||
CR926438.2 | - | 1 - 2 | 122 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |