Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: afg3l1p | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015906.3 | NP_001015906.1 | 16 - 17 | 2165 | 77 | |
GenBank mRNA | BC170788.1 | - | 16 - 17 | 2164 | 77 | |
BC171124.1 | - | 16 - 17 | 2164 | 77 | ||
CR926435.2 | - | 16 - 17 | 2165 | 77 | ||
DR868219.1 | - | 2 - 3 | 398 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |