Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pfdn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015908.2 | NP_001015908.1 | 2 - 3 | 143 | 100 | |
XM_012968347.2 | XP_012823801.1 | 3 - 4 | 231 | 100 | ||
XM_012968348.2 | XP_012823802.1 | 3 - 4 | 328 | 100 | ||
XM_018096247.1 | XP_017951736.1 | 3 - 4 | 154 | 100 | ||
GenBank mRNA | BC135864.1 | - | 3 - 4 | 171 | 100 | |
CR926429.2 | - | 2 - 3 | 143 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |