Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015935.2 | NP_001015935.1 | 5 - 6 | 796 | 69 | |
XM_018095387.1 | XP_017950876.1 | 4 - 5 | 957 | 69 | ||
GenBank mRNA | BC161040.1 | - | 6 - 7 | 850 | 69 | |
CR926369.2 | - | 5 - 6 | 796 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |