Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dusp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016109.2 | NP_001016109.1 | 1 - 2 | 599 | 66 | |
GenBank mRNA | BC170592.1 | - | 1 - 2 | 457 | 66 | |
BC170596.1 | - | 1 - 2 | 457 | 66 | ||
CR762116.2 | - | 1 - 2 | 599 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |