Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016133.2 | NP_001016133.1 | 1 - 2 | 363 | 74 | |
XM_018093063.1 | XP_017948552.1 | 1 - 2 | 1455 | 74 | ||
GenBank mRNA | BC090614.1 | - | 1 - 2 | 363 | 74 | |
CR762041.2 | - | 2 - 3 | 253 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |