Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ugcg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016133.2 | NP_001016133.1 | 4 - 5 | 715 | 99 | |
XM_018093063.1 | XP_017948552.1 | 4 - 5 | 1807 | 99 | ||
GenBank mRNA | BC090614.1 | - | 4 - 5 | 715 | 99 | |
CR762041.2 | - | 5 - 6 | 605 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |