Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufaf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016145.2 | NP_001016145.1 | 4 - 5 | 530 | 103 | |
GenBank mRNA | BC091018.1 | - | 4 - 5 | 530 | 103 | |
CR761995.2 | - | 4 - 5 | 443 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |