Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016148.2 | NP_001016148.1 | 2 - 3 | 141 | 89 | |
XM_012956930.2 | XP_012812384.1 | 3 - 4 | 252 | 89 | ||
XM_012956931.2 | XP_012812385.1 | 3 - 4 | 162 | 89 | ||
GenBank mRNA | CR761990.2 | - | 2 - 3 | 141 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |