Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: samd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016197.2 | NP_001016197.1 | 2 - 3 | 613 | 69 | |
XM_012966564.2 | XP_012822018.1 | 2 - 3 | 1156 | 69 | ||
GenBank mRNA | BC170733.1 | - | 1 - 2 | 582 | 69 | |
BC170735.1 | - | 1 - 2 | 582 | 69 | ||
CR926348.2 | - | 2 - 3 | 613 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |