Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: coq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016225.1 | NP_001016225.1 | 1 - 2 | 109 | 73 | |
GenBank mRNA | CR926300.1 | - | 1 - 2 | 109 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |