Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnal1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016312.2 | NP_001016312.1 | 5 - 6 | 293 | 95 | |
XM_012968391.2 | XP_012823845.1 | 4 - 5 | 536 | 95 | ||
GenBank mRNA | BC158985.1 | - | 4 - 5 | 350 | 95 | |
BC161581.1 | - | 5 - 6 | 307 | 95 | ||
BC170910.1 | - | 5 - 6 | 291 | 95 | ||
BC170912.1 | - | 5 - 6 | 291 | 95 | ||
CR761486.2 | - | 5 - 6 | 293 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |