Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tma7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016374.2 | NP_001016374.1 | 3 - 4 | 168 | 85 | |
GenBank mRNA | BC077652.1 | - | 3 - 4 | 207 | 85 | |
BC135366.1 | - | 3 - 4 | 209 | 85 | ||
CR761261.2 | - | 3 - 4 | 168 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |