Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016428.2 | NP_001016428.1 | 4 - 5 | 496 | 85 | |
GenBank mRNA | BC171075.1 | - | 4 - 5 | 450 | 85 | |
BC171077.1 | - | 4 - 5 | 450 | 85 | ||
CR761059.2 | - | 4 - 5 | 496 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |