Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ucma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016485.2 | NP_001016485.1 | 2 - 3 | 120 | 81 | |
GenBank mRNA | BC135882.1 | - | 2 - 3 | 134 | 81 | |
CR760848.2 | - | 2 - 3 | 120 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |