Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016492.2 | NP_001016492.1 | 4 - 5 | 292 | 88 | |
GenBank mRNA | BC127348.1 | - | 3 - 4 | 286 | 88 | |
BC135545.1 | - | 3 - 4 | 267 | 88 | ||
CR760826.2 | - | 4 - 5 | 292 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |