Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyyr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016493.2 | NP_001016493.1 | 1 - 2 | 212 | 88 | |
GenBank mRNA | CR760825.2 | - | 1 - 2 | 212 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |