Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016523.2 | NP_001016523.1 | 1 - 2 | 153 | 86 | |
GenBank mRNA | BC090592.1 | - | 1 - 2 | 153 | 86 | |
BC122014.1 | - | 1 - 2 | 116 | 86 | ||
BC169172.1 | - | 1 - 2 | 156 | 86 | ||
CR760173.2 | - | 1 - 2 | 124 | 86 | ||
CR926204.2 | - | 1 - 2 | 64 | 86 | ||
CT485744.1 | - | 1 - 2 | 149 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |