Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rassf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016563.1 | NP_001016563.1 | 5 - 6 | 1429 | 97 | |
XM_012961110.2 | XP_012816564.1 | 6 - 7 | 1533 | 97 | ||
XM_012961111.2 | XP_012816565.1 | 5 - 6 | 1515 | 97 | ||
XM_012961112.2 | XP_012816566.1 | 5 - 6 | 1452 | 97 | ||
XM_018093053.1 | XP_017948542.1 | 5 - 6 | 1443 | 97 | ||
GenBank mRNA | CR760685.1 | - | 5 - 6 | 1429 | 97 | |
CU075861.1 | - | 6 - 7 | 1481 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Xenopus tropicalis Probe spans exons |