Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: c1orf35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016609.1 | NP_001016609.1 | 7 - 8 | 643 | 109 | |
XM_012964710.2 | XP_012820164.1 | 8 - 9 | 1140 | 109 | ||
XM_012964711.2 | XP_012820165.1 | 8 - 9 | 914 | 109 | ||
XM_012964712.2 | XP_012820166.1 | 8 - 9 | 1137 | 109 | ||
GenBank mRNA | BC167412.1 | - | 8 - 9 | 700 | 109 | |
CR760531.1 | - | 7 - 8 | 643 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |