Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spcs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016638.2 | NP_001016638.1 | 2 - 3 | 193 | 79 | |
GenBank mRNA | BC155018.1 | - | 221 | 79 | ||
BC157191.1 | - | 2 - 3 | 195 | 79 | ||
BC168790.1 | - | 216 | 79 | |||
CR760420.2 | - | 2 - 3 | 193 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |