Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elovl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016644.2 | NP_001016644.1 | 6 - 7 | 544 | 89 | |
GenBank mRNA | CR760398.2 | - | 6 - 7 | 544 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons |