Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: frzb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016688.2 | NP_001016688.2 | 3 - 4 | 620 | 85 | |
XM_012961122.2 | XP_012816576.1 | 3 - 4 | 787 | 85 | ||
GenBank mRNA | BC158998.1 | - | 3 - 4 | 620 | 85 | |
BC170567.1 | - | 3 - 4 | 518 | 85 | ||
BC170569.1 | - | 3 - 4 | 518 | 85 | ||
CR760192.1 | - | 3 - 4 | 618 | 85 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |