Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016702.2 | NP_001016702.1 | 3 - 4 | 295 | 121 | |
XM_012970435.2 | XP_012825889.1 | 3 - 4 | 356 | 121 | ||
XM_012970436.1 | XP_012825890.1 | 3 - 4 | 292 | 121 | ||
XM_018097109.1 | XP_017952598.1 | 3 - 4 | 384 | 121 | ||
GenBank mRNA | BC091616.1 | - | 2 - 3 | 272 | 121 | |
CR760143.2 | - | 3 - 4 | 295 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |