Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrnp25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016702.2 | NP_001016702.1 | 6 - 7 | 547 | 100 | |
XM_012970435.2 | XP_012825889.1 | 6 - 7 | 608 | 100 | ||
XM_012970436.1 | XP_012825890.1 | 6 - 7 | 544 | 100 | ||
XM_018097109.1 | XP_017952598.1 | 6 - 7 | 636 | 100 | ||
GenBank mRNA | BC091616.1 | - | 5 - 6 | 524 | 100 | |
CR760143.2 | - | 6 - 7 | 547 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |