Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Xt03701555_g1
See other RBM39 GE Assays ›
Gene Symbol:
rbm39
Gene Name:
RNA binding motif protein 39
Gene Aliases:
caper, caperalpha, cc1.3, hcc1, rnpc2
Chromosome Location:
Chr.10: 34295850 - 34311608 on Build UCB_Xtro_10.0
Species:
Xenopus tropicalis
Species Specific ID (Flybase ID):
-
Assay ID
Xt03701555_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: rbm39
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001016788.2
NP_001016788.1
9 - 10
1137
100
XM_012971783.2
XP_012827237.1
9 - 10
1174
100
XM_012971784.2
XP_012827238.1
9 - 10
1171
100
XM_012971785.2
XP_012827239.1
10 - 11
1339
100
XM_012971786.2
XP_012827240.1
9 - 10
1156
100
XM_012971788.2
XP_012827242.1
10 - 11
1287
100
XM_012971789.2
XP_012827243.1
11 - 12
1400
100
XM_012971790.2
XP_012827244.1
10 - 11
1246
100
XM_012971792.2
XP_012827246.1
10 - 11
1271
100
XM_018097748.1
XP_017953237.1
9 - 10
2563
100
XM_018097749.1
XP_017953238.1
10 - 11
1890
100
XM_018097750.1
XP_017953239.1
10 - 11
1838
100
XM_018097751.1
XP_017953240.1
10 - 11
1797
100
XM_018097752.1
XP_017953241.1
11 - 12
1442
100
XM_018097753.1
XP_017953242.1
9 - 10
1174
100
XM_018097754.1
XP_017953243.1
10 - 11
1247
100
XM_018097755.1
XP_017953244.1
9 - 10
1724
100
XM_018097756.1
XP_017953245.1
10 - 11
1340
100
XM_018097757.1
XP_017953246.1
10 - 11
1288
100
XM_018097758.1
XP_017953247.1
9 - 10
2549
100
XM_018097759.1
XP_017953248.1
10 - 11
2588
100
GenBank mRNA
BC093451.1
-
9 - 10
1137
100
CR855673.2
-
9 - 10
1040
100
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Mammalian Gene Collection
Detect Genomic DNA
Xenopus tropicalis
Probe spans exons
Panther Classification:
Molecular Function -
RNA splicing factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0