Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspyl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016811.1 | NP_001016811.1 | 4 - 5 | 1035 | 100 | |
XM_012968444.2 | XP_012823898.1 | 4 - 5 | 1104 | 100 | ||
GenBank mRNA | BC167578.1 | - | 4 - 5 | 1055 | 100 | |
CR855567.1 | - | 4 - 5 | 1035 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |