Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qtrt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016891.2 | NP_001016891.1 | 8 - 9 | 1153 | 57 | |
GenBank mRNA | BC135739.1 | - | 8 - 9 | 1061 | 57 | |
CR848556.2 | - | 8 - 9 | 1153 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |