Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: npm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016938.2 | NP_001016938.1 | 3 - 4 | 254 | 71 | |
XM_012959066.2 | XP_012814520.1 | 3 - 4 | 296 | 71 | ||
XM_012959067.2 | XP_012814521.1 | 3 - 4 | 296 | 71 | ||
GenBank mRNA | BC157168.1 | - | 3 - 4 | 261 | 71 | |
CR762010.2 | - | 3 - 4 | 254 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |