Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016968.2 | NP_001016968.1 | 11 - 12 | 1120 | 63 | |
XM_012961154.2 | XP_012816608.1 | 11 - 12 | 1153 | 63 | ||
GenBank mRNA | CR848466.2 | - | 11 - 12 | 1120 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Xenopus tropicalis Probe spans exons |