Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdcd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017011.1 | NP_001017011.1 | 4 - 5 | 265 | 119 | |
GenBank mRNA | CR762246.1 | - | 4 - 5 | 265 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Xenopus tropicalis Probe spans exons |