Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srd5a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017113.2 | NP_001017113.1 | 2 - 3 | 495 | 100 | |
XM_018093906.1 | XP_017949395.1 | 2 - 3 | 742 | 100 | ||
GenBank mRNA | CR761203.2 | - | 2 - 3 | 495 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |