Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017143.2 | NP_001017143.1 | 2 - 3 | 550 | 92 | |
XM_018093907.1 | XP_017949396.1 | 2 - 3 | 743 | 92 | ||
XM_018093908.1 | XP_017949397.1 | 2 - 3 | 743 | 92 | ||
GenBank mRNA | BC135371.1 | - | 2 - 3 | 557 | 92 | |
BC161042.1 | - | 2 - 3 | 560 | 92 | ||
CR760978.2 | - | 2 - 3 | 550 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |