Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017164.2 | NP_001017164.1 | 2 - 3 | 199 | 102 | |
GenBank mRNA | BC096501.1 | - | 2 - 3 | 199 | 102 | |
CR760800.2 | - | 2 - 3 | 219 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |