Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifi30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017196.2 | NP_001017196.1 | 2 - 3 | 319 | 98 | |
XM_012963420.1 | XP_012818874.1 | 2 - 3 | 489 | 98 | ||
GenBank mRNA | CR760545.2 | - | 2 - 3 | 319 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Xenopus tropicalis Probe spans exons |