Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017205.2 | NP_001017205.1 | 2 - 3 | 381 | 104 | |
GenBank mRNA | BC135154.1 | - | 2 - 3 | 392 | 104 | |
CR760502.2 | - | 2 - 3 | 381 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |