Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prxl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017220.2 | NP_001017220.1 | 6 - 7 | 597 | 103 | |
XM_012966658.2 | XP_012822112.1 | 6 - 7 | 621 | 103 | ||
GenBank mRNA | CR760360.2 | - | 6 - 7 | 597 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Xenopus tropicalis Probe spans exons |