Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufb5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017303.2 | NP_001017303.1 | 3 - 4 | 281 | 77 | |
XM_018093917.1 | XP_017949406.1 | 3 - 4 | 259 | 77 | ||
GenBank mRNA | BC152034.1 | - | 3 - 4 | 294 | 77 | |
CR855571.2 | - | 3 - 4 | 281 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |