Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zdhhc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017307.2 | NP_001017307.1 | 1 - 2 | 150 | 99 | |
GenBank mRNA | BC170884.1 | - | 1 - 2 | 129 | 99 | |
BC170886.1 | - | 1 - 2 | 129 | 99 | ||
CR855529.2 | - | 1 - 2 | 150 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |